BioBlock™ 96-Well Deep Well Plates, Simport Scientific
Supplier: Simport Scientific
T110-5 T110-6 T110-10 T110-5B T110-5P T110-5Y T110-10B T110-10G T110-10P T110-10Y T110-5 T110-6 T110-10G
CA48648-151CS 283.17 CAD
CA48648-151 CA48648-152 CA48648-153 CA11028-452 CA11028-454 CA11028-456 CA11028-458 CA11028-462 CA11028-464 CA11028-466 CA11028-468 89499-340 89499-350 89499-360
BioBlock™ 96-Well Deep Well Plates, Simport Scientific
Microplates Deep Well Microplates
BioBlock™ deep well plates conform to the SBS standard footprint and 96-well microtiter plates
The polypropylene T110-5 and T110-10 series are available in a choice of Natural, Blue, Green, Pink, or Yellow colors.
Microplates are compatible with all leading robotic sample processors and automated liquid handling systems. The polypropylene T110-5 and T110-10 series are DMSO resistant and withstand centrifugation up to 6000 g. Polystyrene model T110-6 is not DMSO resistant but will withstand centrifugation up to 3000g. All polypropylene microplates offer an operating temperature range of –196 to 121 °C. Microplates are available in 1.0 and 2.0 mL volumes. All plates feature round bottoms with an alphanumeric grid to facilitate sample identification.
Ordering information: Mat covers are sold separately. See items 89499-396 and 89499-398.