Grip Racks with Tube Ejector, Globe Scientific
Supplier: Globe Scientific
456925 456926
89498-058EA 84.55 CAD
89498-058 89498-060
Grip Racks with Tube Ejector, Globe Scientific
Racks Tube Racks
These specially designed test tube racks feature rubber grips that hold test tubes in place when decanting liquid.
  • Produced from polyoxymethylene (POM)
  • Holds various size tubes up to 15 mm diameter
  • 50-place rack (5×10 configuration)
  • Dimensions: 260×140×82 mm (10×5.5×3")
  • Autoclavable
The racks also feature a unique tube ejector that allows the user to easily eject all tubes into a waste receptacle.