Low Profile Grip Racks, Globe Scientific
Supplier: Globe Scientific
456920 456921
89498-054EA 62.65 CAD
89498-054 89498-056
Low Profile Grip Racks, Globe Scientific
Racks Tube Racks
These specially designed test tube racks feature rubber grips that hold test tubes in place when decanting liquid.
  • Produced from polyoxymethylene (POM)
  • Holds various size tubes up to 17 mm diameter
  • Low profile design allows for easy viewing of labels on tubes
  • 50-place rack (5×10 configuration)
  • Dimensions: 254×140×51 mm (10×5.5×2")
  • Autoclavable