Snap Racks for 30 mm Tubes, Globe Scientific
Supplier: Globe Scientific
456803 456804 456806 456810
89498-046EA 68.25 CAD
89498-046 89498-048 89498-050 89498-052
Snap Racks for 30 mm Tubes, Globe Scientific
Racks Tube Racks
These innovative polypropylene test tube racks are shipped flat in order to reduce shelf space.
  • Polypropylene (PP)
  • Autoclavable
  • Non-floating in water baths
  • Stackable when empty
  • Ships flat for ease of storage
Snap racks feature molded alphanumeric markings to enable quick and easy test tube identification. Available in assorted colors. Snap racks are non-floating in water baths.
They simply snap together for use. The special design allows all samples to be incubated at the same temperature. The racks can be used in freezers without deforming orMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable