Snap Racks for 25 mm Tubes, Globe Scientific
Supplier: Globe Scientific
456703 456704 456706 456710
89498-038EA 62.38 CAD
89498-038 89498-040 89498-042 89498-044
Snap Racks for 25 mm Tubes, Globe Scientific
Racks Tube Racks
These innovative polypropylene test tube racks are shipped flat in order to reduce shelf space.
  • Polypropylene (PP)
  • Autoclavable
  • Non-floating in water baths
  • Stackable when empty
  • Ships flat for ease of storage
Snap racks feature molded alphanumeric markings to enable quick and easy test tube identification. Available in assorted colors. Snap racks are non-floating in water baths.
They simply snap together for use. The special design allows all samples to be incubated at the same temperature. The racks can be used in freezers without deforming or becoming brittle. They will not rot, rust, or deform.