Snap Racks for Microcentrifuge Tubes, Globe Scientific
Supplier: Globe Scientific
456303 456304 456306 456310
89498-004EA 52.73 CAD
89498-004 89498-006 89498-008 89498-010
Snap Racks for Microcentrifuge Tubes, Globe Scientific
Racks Tube Racks
These innovative polypropylene test tube racks are shipped flat in order to reduce shelf space.
  • Polypropylene (PP)
  • Autoclavable
  • Non-floating in water baths
  • Stackable when empty
  • Ships flat for ease of storage
Snap racks feature molded alphanumeric markings to enable quick and easy test tube identification. Available in assorted colors. Snap racks are non-floating in water baths.
They simply snap together for use. The special design allows all samples to be incubated at the same temperature. The racks can be used in freezers without deforming orMore Product Information