Polystyrene and Polypropylene Test Tubes, 12×75 mm, Globe Scientific
Supplier: Globe Scientific
110410 110405 110406 110410I 110410LB 110412 110413 110414 110428 110438 110441 110441AM 110442 110443 110444 110446 110416 110440 110409
89209-796PK 90.16 CAD
89209-796 89497-794 89497-796 89497-798 89497-800 89497-802 89497-804 89497-806 89497-810 89497-812 89497-814 89497-816 89497-818 89497-820 89497-822 10014-224 10014-242 10014-244 10014-252
Polystyrene and Polypropylene Test Tubes, 12×75 mm, Globe Scientific
Tubes Test Tubes
These tubes are molded from either premium-grade polystyrene for clarity, or polypropylene for impact strength and stability to common acids, alkalis, and solvents.
Ordering information: Unless otherwise specified, test tubes are supplied without caps.