Polystyrene and Polypropylene Test Tubes, 17×100 mm, Globe Scientific
Supplier: Globe Scientific
110158 110178 110182 110184 110186
89497-782CS 430.53 CAD
89497-782 89497-784 89497-788 89497-790 89497-792
Polystyrene and Polypropylene Test Tubes, 17×100 mm, Globe Scientific
Tubes Test Tubes
These tubes are molded either from premium-grade polystyrene for clarity, or polypropylene for impact strength and stability to common acids, alkalis and solvents.
Caps are made of polyethylene and are not autoclavable.