CF™ Dye Streptavidin and Monoclonal Mouse Anti-Biotin Conjugates, Biotium
Supplier: Biotium
29036 29037 20203 20203-1 20204 20204-1 20205 20206 20206-1 20207 20207-1 29031 29032 29033 29034 29035 29038 29039 29044-1ML 29044-200UL 29030 20367-250UL 20367-50UL
89138-874EA 470.54 CAD
89138-874 89138-876 97065-632 97065-634 97065-636 97065-638 97065-640 97065-644 97065-646 97065-648 97065-650 97065-740 97065-742 97065-744 97065-746 97065-748 97065-750 97065-752 89411-136 89411-548 89427-112 89427-246 89427-248
CF™ Dye Streptavidin and Monoclonal Mouse Anti-Biotin Conjugates, Biotium
Protein Linking and Labeling Systems
Biotium offers a variety of streptavidin and anti-biotin antibody conjugates featuring our series of CF dyes
Streptavidin conjugates are typically used as secondary reagents to detect biotinylated probes such as primary antibodies for flow cytometry, Western blotting, immunofluorescence staining and other applications.
Monoclonal mouse anti-biotin antibody reacts with biotin conjugated to antibodies or other proteins, and may show less non-specific binding to cells or tissues compared to streptavidin conjugates.
CF dyes are superior to other fluorescent dyes for protein labeling due to advantages in brightness, photostability, and water solubility.
Ordering information: Streptavidin conjugates are supplied in lyophilized solid format. After reconstitution with 0.5 mL PBS, streptavidin concentration is 2 mg/mL with 0.01% sodium azide. Monoclonal mouse anti-biotin antibody conjugates are supplied at 2 mg/mL in PBS pH 7.4 containing 50% glycerol, 2 mg/mL BSA (IgG-free and protease-free), and 0.05% sodium azide.