EverBrite™ Fluorescence Antifade Mounting Media and CoverGrip™ Coverslip Sealant, Biotium
Supplier: Biotium
23001 23002 23005 23005-1 23003 23004
89411-104EA 337.31 CAD
89411-104 89411-106 89411-108 89411-134 89427-068 89427-070
EverBrite™ Fluorescence Antifade Mounting Media and CoverGrip™ Coverslip Sealant, Biotium
Stains Microscopy and staining reagents
EverBrite™ Mounting Media are anti-fade mountants for preserving fluorescence during microscopy
EverBrite™ Mounting Medium is a wet-set medium, while EverBrite™ Hardset Mounting Medium cures overnight to form a hard, permanent seal with the coverslip, eliminating the need to seal coverslip edges with nail polish or other sealants. Both formulations are available with DAPI for blue fluorescent nuclear counterstaining without an additional staining step.
CoverGrip™ Coverslip Sealant is an organic adhesive medium formulated specifically for sealing the edges of wet-mounted coverslips. Unlike nail polish, CoverGrip™ does not contain alcohol, which can diffuse into aqueous mounting medium and affect sample fluorescence. CoverGrip™ is made with limonene, a pleasant-smelling natural hydrocarbon made from citrus peels that is more environmentally friendly and less toxic than other organic solvents. CoverGrip™ is supplied in a 15 mL brush cap bottle for ease of application, or in 100 mL dropper-top refill bottles.
EverBrite™ Mounting Media prevent rapid photobleaching of fluorescein, rhodamine, and cyanine based dyes and are optimally formulated for use with Biotum’s CF™ dye conjugates. Unlike Vectashield®, EverBrite™ mounting media is compatible with Cy® dyes such as Cy®2, Cy®3, and Cy®5.