dUTP CF® Dye Conjugates, Biotium
Supplier: Biotium
CF® dye-dUTP can be used for TUNEL assay, microarray, or to synthesize labeled DNA probes for in situ hybridization and nucleic acid blotting applications.
- For labeling fluorescent DNA probes
- Detect by fluorescence microscopy, flow cytometry, or in gel
- Compatible with fixed cells or tissue sections
- Choice of 8 CF® dye colors from blue to near infrared
- Biotium’s CF® dyes have superior brightness, photostability, and are water-soluble
Biotium’s next-generation CF® dyes were designed to be highly water-soluble with advantages in brightness and photostability compared to other fluorescent dyes. CF® dye dUTP conjugates are supplied in lyophilized form.
Caution: For PCR applications, Taq polymerase should be used with dUTP conjugates, because dUTP inhibits archaeal polymerases such as Pfu and Vent®.