CF™ Dye Bovine Serum Albumin Conjugates, Biotium
Supplier: Biotium
Fluorescent bovine serum albumin (BSA) conjugates are commonly used as tracers in applications where physical dimensions are important
CF dye technology is covered by pending U.S. and international patents.
The conjugates can also be used for quantitative studies of electroporation, measurement of plasma volume in rats, and endocytosis studies. CF™ dye BSA conjugates are labeled with Biotium’s next-generation fluorescent CF™ dyes, which offer advantages in brightness and photostability compared to other fluorescent dyes.