CytoSep™ Cytology Funnel Chambers for the Hettich Cyto-System, Simport Scientific
Supplier: Simport Scientific
M966-1 M966-2 M966-4 M966-8 M966FW M966FW8 M966-1 M966-2 M966-4 M966-8 M966FW M966FW8
89305-124EA 165.06 CAD
89305-124 89305-126 89305-128 89305-130 89305-132 89305-134 CA89305-124 CA89305-126 CA89305-128 CA89305-130 CA89305-132 CA89305-134
CytoSep™ Cytology Funnel Chambers for the Hettich Cyto-System, Simport Scientific
Funnels Cytology Funnels
The Simport CytoSep™ Funnel Chambers for the Hettich Cyto-System optimize lab throughput with multiple funnel
Plastic components made of polypropylene.
These funnels are perfect for every application such as CSF, Viral infections, Bronchial secretions and come in four sizes from 1 ml to 8 ml. The 1 ml chamber is for small-volume samples of low cell contents. Both 2 and 4 ml chambers are suggested for cell-rich samples, e.g. pleura, ascites and bronchial washings while the 8 ml chamber is perfect for large-volume samples such as urine.
Certifications: CE Marked.