CritiCore Cleanroom Frocks with ESD Grid
Supplier: VWR International
89495-836 89495-838 89495-840 89495-842 89495-844 89495-846 89495-848 89495-850 89495-852 89495-854 89495-856 89495-858 89495-860 89495-862 89495-864 89495-866 89495-868 89495-870
89495-836EA 156.25 CAD
89495-836 89495-838 89495-840 89495-842 89495-844 89495-846 89495-848 89495-850 89495-852 89495-854 89495-856 89495-858 89495-860 89495-862 89495-864 89495-866 89495-868 89495-870
CritiCore Cleanroom Frocks with ESD Grid
Workwear
Reusable zippered frocks are designed for controlled environments and less critical cleanroom applications. Made with durable ESD Grid Fabric, they are engineered for electrostatic dissipation with a 1% carbon grid, and also feature knit cuffs with 5% carbon Belltron fiber. Applications include micro-electronics and semi-conductor manufacturing.
  • Zippered, durable, and reusable
  • For micro-electronics and semi-conductor manufacturing
  • All seams are laser cut or pre-serged, silicon-free thread
Garments have a mandarin collar with two snaps at top, a wide-pull zipper, and a single snap below at bottom
Available in white and navy.
Assembled with double-needle felled seams for added durability and particle migration prevention. All seams are laser-cut or pre-serged, and thread is silicon-free.
Ordering information: Custom order product with extended lead time. Contact your Avantor sales representative for more information.