Cryostore™ Storage Boxes for Cryogenic Vials, Simport Scientific
Supplier: Simport Scientific
T314-225B T314-225G T314-225R T314-225Y T314-481B T314-481G T314-481R T314-481Y T314-2100Y T314-581B T314-581G T314-581R T314-581Y T314-281B T314-281R T314-2100B T314-2100G T314-2100R T314-2100B T314-2100G T314-2100R T314-2100Y T314-225B T314-225G T314-225R T314-225Y T314-281B T314-281G T314-281R T314-281Y T314-481B T314-481G T314-481R T314-481Y T314-581B T314-581G T314-581R T314-581Y T314-542B T314-542G T314-542R T314-542Y T314-225B
CA11006-966CS 659.41 CAD
CA11006-966 CA11006-968 CA11006-970 CA11006-972 CA11006-974 CA11006-976 CA11006-978 CA11006-980 CA11007-768 CA48648-110 CA48648-111 CA48648-112 CA48648-113 CA48648-106 CA48648-108 CA11008-454 CA11008-456 CA11008-458 89492-666 89492-668 89492-670 89492-672 89492-674 89492-676 89492-678 89492-680 89492-682 89492-684 89492-686 89492-688 89492-690 89492-692 89492-694 89492-696 89492-698 89492-700 89492-702 89492-704 CA11028-524 CA11028-526 CA11028-528 CA11028-530 490014-848
Cryostore™ Storage Boxes for Cryogenic Vials, Simport Scientific
Boxes Cryoboxes
Made of extra strong polycarbonate, these durable cryogenic storage boxes are designed to be used at temperatures between -196 °C and +121 °C and are autoclavable at 120 °C, 15 psig (1 bar) for 20 minutes
Colored plastic grids separate the cover from the base on the 25, 42 and 81-place boxes. Those made to accept 100 tubes have a colored base instead of a grid.
A transparent cover allows the user to see the contents of the box, and is keyed to the base to prevent misalignment. Printed with a series of squares (numbered from 1 to 25, 1 to 42, 1 to 81, or 1 to 100), surface accepts writing with markers, facilitating inventory control.