Cryogenic Vials, Internal or External Threaded, Simport Scientific
Supplier: Simport Scientific
T308-3A T308-1A T308-2A T308-2 T308-4A T308-5A T311-1 T311-2 T311-3 T311-5 T310-5A T310-10A T310-4A T310-3A T310-2 T310-2A T309-5A T309-4A T309-3A T309-2A T309-2 T309-1A T311-4A T311-4 T309-1A T309-2A T309-3A T309-5A T310-3A T301-1 T301-2 T301-3 T301-1 T301-2 T301-3 T301-5 T308-1A T308-2 T308-2A T308-4A T308-5A T311-1 T311-4A T301-5 T301-4 T301-4A T301-4 T301-4A
CA11006-292CS 2065.39 CAD
CA11006-292 CA11006-956 CA11006-958 CA11006-960 CA11006-962 CA11006-964 CA60872-110 CA60872-120 CA60872-130 CA60872-150 CA73960-370 CA73960-371 CA73960-372 CA73960-374 CA73960-376 CA73960-378 CA73960-392 CA73960-394 CA73960-396 CA73960-398 CA73960-400 CA73960-402 CA73960-416 CA73960-418 89108-138 89108-142 89108-144 89108-148 89108-150 CA11008-448 CA11008-450 CA11008-452 89492-644 89492-646 89492-648 89492-650 89492-652 89492-654 89492-656 89492-658 89492-660 89492-662 89492-664 CA11028-522 10062-820 10062-822 CA10120-198 CA10120-200
Cryogenic Vials, Internal or External Threaded, Simport Scientific
Vials
Cryogenic Vials are designed for storing biological material, human or animal cells at temperatures as low as −196 °C.
  • Large white marking area and printed graduations
  • Model T310-10A has a polyethylene screw cap
Closures and tubes are both made of polypropylene having the same coefficient of expansion which further enhances the leakproof qualities of these vials at various temperatures.
Certifications: Certified Rnase-, DNase-, Pyrogen- and DNA-free. CE marked.
Ordering information: Tubes can be color coded by the use of CAPINSERT Accessory.