Storage Cane for Cryovial® Tubes or Cryogenic Vials, Simport Scientific
Supplier: Simport
T313
CA48648-500CS 103.58 CAD
CA48648-500
Storage Cane for Cryovial® Tubes or Cryogenic Vials, Simport Scientific
Cryogenic Straws
For storage of up to five 1.2 or 2 mL Simport Cryovial® tubes in liquid nitrogen containers such as Dewar flasks.
Made of aluminum.