SP Bel-Art Biohazard Incinerator Cartons, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F13205-0002 F13205-0001
70680-350PK 172.85 CAD
70680-350 70680-352
SP Bel-Art Biohazard Incinerator Cartons, Bel-Art Products, a part of SP
Boxes Waste Disposal Boxes
This corrugated cardboard receptacle is perfect for containing biohazardous waste slated for incineration.
  • Corrugated cardboard construction
  • Contains biohazardous waste without exposing users
  • Bright, large biohazard graphics printed on side
  • Optional reusable cover
When the box is full, pull the flap to a closed position and the entire unit can be placed in the incinerator. Tabs along the top edge keep a clear 0.038 mm (1.5 mil) thick polyethylene biohazard bag from slipping out of place and the snug-fitting lid has a flap for easily inserting waste material. Printed with large, bright, biohazard graphics and instructions in four languages (English, Spanish, French, German), it is available in sizes to suit benchtop or floor use.
An optional, reusable, sanitary cover (89088-048) for floor models is sold separately. Dishwasher-safe and autoclave-safe, the reusable Disposal Carton Cover opens quickly and easily by pressing the built-in release tab. No batteries are required. The cover fits any disposal carton that has a 30.5 x 30.5 cm (12 x12") opening, and features a smooth 10.1 x 10.1 cm (4 x 4") area on top for applying labels. The center panel is red to match the printing on the Biohazard disposal cartons, providing fast visual recognition of the waste stream. Compared to the disposable cardboard lids supplied with the cartons, the Disposal Carton Cover provides a wider opening, 20.3 x 24.1 cm (8 x 9.5"), permitting safer and more sanitary deposit of wastes into the container. Store the disposable cardboard lid on the bottom of the carton, and when carton is full, simply remove the reusable Disposal Carton Cover, replace with the cardboard lid, and dispose of the carton in the usual fashion. Wash the Disposal Carton Cover as needed, and it’s ready for re-use. Autoclavable at 121 °C (250 °F).
Ordering information: Packaged flat to save space and shipping costs; carton and inner bag can be assembled in seconds.