Distillation Receiver, Vertical Top, Ace Glass
Supplier: Ace Glass
9401-05 9401-30
89074-470EA 187.32 CAD
89074-470 89074-472
Distillation Receiver, Vertical Top, Ace Glass
Distillation Components Distillation Receivers
This distillation receiver is for use where separate fractions are desired without disturbing the distillation pressure
Has [ST] 14/20 joints. Receiver flasks are 5 mL. Hose connections are for use with 3/8" or 5/16" I.D. tubing. Fabricated from borosilicate glass.
Ordering information: Complete [ST] 14/20 size includes the following items: vacuum adapter (89074-470), distributor (89074-460), four 5 mL flasks (89074-584), and five Dupont Delrin joint clips/clamps (89059-608). For PTFE-clad version, see 89074-462. For [ST] 24/40 version (NOT PTFE-clad), see 89055-730.