Corning® PureCoat™ Amine and Carboxyl Microplates, Corning
Supplier: Corning
356717 356719 354717 354719 356717 356719
CA11006-198CS 2416.26 CAD
CA11006-198 CA11006-200 CA95059-884 CA95059-886 CA95059-904 CA95059-906
Corning® PureCoat™ Amine and Carboxyl Microplates, Corning
Microplates Cell Culture Plates
Corning PureCoat Amine (positively charged) and Carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.