Cell culture dishes, amine and carboxyl coated, Corning® PureCoat™
Supplier: Corning
354732 356732
CA11006-186CS 171.79 CAD
CA11006-186 CA11006-206
Cell culture dishes, amine and carboxyl coated, Corning® PureCoat™
Culture Dishes Cell Culture Dishes
PS. Corning® PureCoat™ surfaces are a novel family of chemically-defined, animal-free cell culture surfaces designed to enhance cell performance. A proprietary thin-film coating technology produces a uniform, functionalised surface that provides a highly controlled environment for cell culture applications.
  • Available as amine (positively charged) or carboxyl (negatively charged) surface chemistries
  • Optimised for culture in low serum or no serum conditions
  • Enhanced freeze-thaw recovery of cryopreserved cells
  • Improved differentiation of cells
  • Culture of many
More Product Information