Corning® Cell Recovery Solution, Corning
Supplier: Corning
Corning®
354270 354253 354253
CACB354253EA 106.35 CAD
47743-696 CACB354253 76332-050
Corning® Cell Recovery Solution, Corning
Cell Culture Supplements and Additives
Corning® Cell Recovery Solution allows for the recovery of cells cultured on Corning® Matrigel® Basement Membrane Matrix for subsequent biochemical analysis. Corning® Cell Recovery Solution depolymerises Corning® Matrigel® Matrix gels without enzymatic digests and lengthy incubation periods at high temperatures. Cells are released without damage thereby avoiding biochemical changes during incubation and digestion of extracellular portions of cell-surface receptors and adhesion molecules.
  • Non-enzymatic proprietary solution
  • Tested for ability to depolymerise a 1 mm thick layer of gelled BD Matrigel™ matrix after one hour at 2 to 8 °C
  • Filtered and tested and found negative for bacteria, fungi, and mycoplasma
  • Stable for at least three months at 2 to 8 °C