Corning® Cell and Tissue Adhesive, Corning
Supplier: Corning
Corning®
354241 354242 354240 354242 354241 354240
CACB354240EA 361.74 CAD
CACB354240 47743-684 47743-686 CACB-354242 CACB354241 47743-726
Corning® Cell and Tissue Adhesive, Corning
Adhesives
Corning® Cell-Tak™ Cell and Tissue Adhesive, is used to attach cells or tissue sections to many types of surfaces, including plastic, glass, metal, FEP Polymer, and biological materials.