WaterPro® PS HPLC/UF Hybrid, Labconco
Supplier: Labconco
9000722 9000732 9000742 9000762 9000772 9000782
89496-552EA 19323.4 CAD
89496-552 89496-554 89496-556 89496-558 89496-560 89496-562
WaterPro® PS HPLC/UF Hybrid, Labconco
Water Purification Systems Deionizers
WaterPro PS Polishing Stations deliver Type I, 18.2 megohm-cm water from the dispensing valve or optional dispensing gun (refer to descriptions for models that include gun) at a typical rate of 1.8 liters per minute depending on feedwater flow rate and pressure, and installed purification modules.
The control panel includes a timed dispense feature that allows unattended dispensing of pure water without risk of overfilling. The dispenser shuts off automatically once user-set time has elapsed. Labconco has designed WaterPro PS Stations with the right combinations of purificationMore Product Information