WaterPro® PS/UF Life Science Model, Labconco
Supplier: Labconco
9000720 9000730 9000740 9000760 9000770 9000780 9000721 9000731 9000741 9000761 9000771 9000781
89496-528EA 15815.8 CAD
89496-528 89496-530 89496-532 89496-534 89496-536 89496-538 89496-540 89496-542 89496-544 89496-546 89496-548 89496-550
WaterPro® PS/UF Life Science Model, Labconco
Water Purification Systems Deionizers
WaterPro PS Polishing Stations deliver Type I, 18.2 megohm-cm water from the dispensing valve or optional dispensing gun (refer to descriptions for models that include gun) at a typical rate of 1.8 liters per minute depending on feedwater flow rate and pressure, and installed purification modules.
The control panel includes a timed dispense feature that allows unattended dispensing of pure water without risk of overfilling. The dispenser shuts off automatically once user-set time has elapsed. Labconco has designed WaterPro PS Stations with the right combinations of purification technologies to meet your needs.
The WaterPro PS/UF Life Science model includes four bowls constructed of polypropylene for one carbon and three deionization cartridges (all cartridges sold separately).