Cylinder Round Bottles, High Density Polyethylene, Narrow Mouth, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
221163 221164
16300-039CS 280.41 CAD
16300-039 16300-040
Cylinder Round Bottles, High Density Polyethylene, Narrow Mouth, WHEATON®, DWK Life Sciences
Bottles Plastic Bottles
Round, narrow mouth plastic bottles are convenience packed with foamed polyethylene lined polypropylene closures attached.
White HDPE helps protect light sensitive samples.
The narrow mouth is ideal for liquids.
Ordering information: Bottles are packaged 12 per pack and 6 packs per case.