IL Analyzer Supplies, Globe Scientific
Supplier: Globe Scientific
5150
60820-040CS 935.09 CAD
60820-040
IL Analyzer Supplies, Globe Scientific
Cuvettes Specialty Cuvettes
Globe Scientific's 20-place cuvette rotors are designed for use with the IL ACL® models: 100, 200, 300, 1000, 2000, 3000, 6000 & 7000.
The cuvette rotors are packaged 25 per sleeve, 4 packs per case.
Please note: Not for use on the ACL 9000 & 10000.