Centrifuge Tubes, 12 ml, Plain Top, Globe Scientific
Supplier: Globe Scientific
112010-500 112040
60819-980CS 166.4 CAD
60819-980 10022-254
Centrifuge Tubes, 12 ml, Plain Top, Globe Scientific
Tubes Centrifuge Tubes
Globe's 12 ml general-purpose centrifuge tubes are offered in polystyrene (PS) and polypropylene (PP).
The tubes are graduated at 0.5, 1, 2.5, 5, and 10 ml.