Plain Top Centrifuge Tubes, 15 ml, Globe Scientific
Supplier: Globe Scientific
6260 6266 6261 6262
60819-972CS 508.1 CAD
60819-972 10022-238 10022-240 10022-256
Plain Top Centrifuge Tubes, 15 ml, Globe Scientific
Tubes Centrifuge Tubes
These general-purpose plain top centrifuge tubes are available in polypropylene (PP) or polystyrene (PS).
The tubes are graduated from 0.5 to 15 ml in 0.5 ml increments.
Packaging: Available in bulk or packaged in convenient dispenser boxes.