Media Lab Bottles, Amber, Graduated, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
219865 219845 219855
16149-170CS 1754.3 CAD
16149-170 16159-670 16159-674
Media Lab Bottles, Amber, Graduated, WHEATON®, DWK Life Sciences
Bottles Cell Culture Bottles
Wheaton media bottles are manufactured from Wheaton 400 borosilicate glass.
  • Media lab bottles have 25 ml graduations and a writing patch
The glass conforms to USP Type I requirements and has a low alkali content to help prevent changes in pH. This also helps to maintain the purity of its contents. The pour lip and sloped shoulders facilitate pouring and cleaning. Pre-attached black phenolic screw caps with liner are attached to each bottle.