VWR® Static Shielding Reclosable Bags
Supplier: VWR International
89071-966 89071-978
89071-966CS 43.26 CAD
89071-966 89071-978
VWR® Static Shielding Reclosable Bags
Bags Storage Bags
VWR® static-shielding reclosable bags provide total Faraday Cage protection and visibility for highly sensitive electronic components.
  • Reclosable design
  • Polycarbonate safe
  • Contains no amines, amides or N-Octanic acid
  • Static shielding notice stamped in yellow on each bag
Static Shielding bags guard against static charges from both inside and outside, with three layers of anti-static protection. Rugged 3 mm zip top bag are 50% transparent for easy product identification. This bag has a buried metal-in construction; a conductive aluminum shield layer is buried between two static dissipative polymeric plastic layers.
Certifications: Meets the electrical and physical requirements of ANSI/ESD S541, EIA 625 and are ANSI/ESD S20.20 program compliant. Tested per MIL-STD-3010 4046 and ANSI/ESD STM11.11.