Reagent Bottles, Narrow Mouth, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W215235 W215255 215259 215257 215237 215239 215243 215241 215240
14216-238CS 347.19 CAD
14216-238 14216-240 16158-313 16158-317 16262-042 16262-044 74830-122 74830-124 74830-126
Reagent Bottles, Narrow Mouth, WHEATON®, DWK Life Sciences
Bottles Glass Bottles
Wheaton reagent bottles are manufactured from USP Type I borosilicate glass.
  • Reagent bottles and stoppers are autoclavable
Each bottle comes with a ground glass stopper and features a no drip pour lip. The narrow mouth is ideal for liquids. The amber bottles designed for use with light sensitive samples during storage.