Tite-Rite™ Containers, Globe Scientific
Supplier: Globe Scientific
6525 6520 6522 6524 6527 6518
10030-350CS 696.57 CAD
10030-350 59591-205 10030-306 10030-356 10030-358 10030-352
Tite-Rite™ Containers, Globe Scientific
Sample Containers General Purpose Containers
Tite-Rite™ containers provide the ultimate convenience and reliability in leak-tight storage and liquid transport
Manufactured from polypropylene, these conatiners are autoclavable at 121°C (250°F) for 20 minutes (without caps). Containers are 95kPa compliant. Caps are manufactured from polyethylene.
The specially designed caps contain an inner liner designed to compress against the uppermost walls of the container, ensuring an air-tight seal. The containers are ideal for use in hospital pneumatic tube systems.