WHEATON® Vial Racks, DWK Life Sciences
Supplier: DWK Life Sciences
868808 985800 868804 868806 868805 868810 985750
14007-178CS 658.11 CAD
14007-178 66022-478 66022-523 66022-525 66022-556 66022-569 66022-580
WHEATON® Vial Racks, DWK Life Sciences
Racks Cryogenic Vial Racks
Manufactured from polypropylene, vial racks are easy to clean and autoclavable
Each rack features alphanumeric indexing to simplify identification and selection. The racks are sturdy and stackable, even when vials are in place.
Ordering information: When selecting the proper rack, the vial O.D. cannot exceed the well I.D. dimension.
Order Now
SPECIFICATIONS
Environmentally Preferable