Snap Caps for Blood Drawing Tubes, Globe Scientific
Supplier: Globe Scientific
113146C 113148C
89209-806PK 73.29 CAD
89209-806 89209-808
Snap Caps for Blood Drawing Tubes, Globe Scientific
Closures Test/Sample Tube Closures
Snap-on closures are an effective way to recap blood drawing tubes, glass test tubes, and plastic culture tubes
They are precision molded from low-density polyethylene (LDPE) and maintain a tight seal during centrifugation, agitation, and refrigeration procedures.