VENT® DNA Polymerase, New England Biolabs
Supplier: New England Biolabs (NEB)
M0254L
M0254S
CA101228-290EA
504
CAD
CA101228-290
CA101228-292
VENT® DNA Polymerase, New England Biolabs
Nucleic Acid Reagents
End-point PCR Enzymes and Kits
VentR DNA Polymerase was the first high-fidelity thermophilic DNA polymerase available for PCR
The fidelity is 5-15-fold higher than that observed for Taq DNA Polymerase, and is derived in part from an integral 3´→ 5´ proofreading exonuclease activity in Vent DNA Polymerase. Greater than 90% activity remains following a 1 hour incubation at 95°C.