VENT® (exo-) DNA Polymerase, New England Biolabs
Supplier: New England Biolabs (NEB)
M0257L
M0257S
CA101228-298EA
504
CAD
CA101228-298
CA101228-300
VENT® (exo-) DNA Polymerase, New England Biolabs
Nucleic Acid Reagents
End-point PCR Enzymes and Kits
VentR (exo-) DNA Polymerase has been genetically engineered to eliminate the 3´→ 5´ proofreading exonuclease activity associated with VentR DNA Polymerase
This is the preferred form for high-temperature dideoxy sequencing reactions and for high yield primer extension reactions. The fidelity of polymerization by this form is reduced to a level about 2-fold higher than that of Taq DNA Polymerase.