USER
Supplier: New England Biolabs (NEB)
M5505L M5505S
CA101228-398EA 511 CAD
CA101228-398 CA101228-400
USER
Enzymes
USER (Uracil-Specific Excision Reagent) Enzyme generates a single nucleotide gap at the location of a uracil
USER Enzyme is a mixture of Uracil DNA glycosylase (UDG) and the DNA glycosylase-lyase Endonuclease VIII. UDG catalyses the excision of a uracil base, forming an abasic (apyrimidinic) site while leaving the phosphodiester backbone intact. The lyase activity of Endonuclease VIII breaks the phosphodiester backbone at the 3´ and 5´ sides of the abasic site so that base-free deoxyribose is released.