Uracil-DNA Glycosylase
Supplier: New England Biolabs (NEB)
M0280L
M0280S
CA101228-336EA
518
CAD
CA101228-336
CA101228-338
Uracil-DNA Glycosylase
Enzymes
E.coli Uracil-DNA Glycosylase (UDG) catalyses the release of free uracil from uracil-containing DNA. UDG efficiently hydrolyzes uracil from single-stranded or double-stranded DNA, but not from oligomers (6 or fewer bases).