Uracil Glycosylase Inhibitor
Supplier: New England Biolabs (NEB)
M0281L
M0281S
CA101228-340EA
505
CAD
CA101228-340
CA101228-342
Uracil Glycosylase Inhibitor
Enzymes
The Uracil Glycosylase Inhibitor (UGI) of Bacillus subtilis bacteriophage PBS1 is a small protein (9.5 kDa) which inhibits E.coli uracil-DNA glycosylase (UDG) as well as UDG from other species. Inhibition of UDG occurs by reversible protein binding with a 1:1 UGD:UGI stoichiometry. UGI is capable of dissociating UDG-DNA complexes.