Thermostable Inorganic Pyrophosphatase
Supplier: New England Biolabs (NEB)
M0296L M0296S
CA101228-360EA 505 CAD
CA101228-360 CA101228-362
Thermostable Inorganic Pyrophosphatase
Enzymes
Inorganic pyrophosphatase (PPase) catalyzes the hydrolysis of inorganic pyrophosphate to form orthophosphate: P2O7-4 + H2O -> 2HPO4-2.