THERMINATOR™ DNA Polymerase, New England Biolabs
Supplier: New England Biolabs (NEB)
M0261L M0261S
CA101227-268EA 727 CAD
CA101227-268 CA101227-270
THERMINATOR™ DNA Polymerase, New England Biolabs
Nucleic Acid Reagents Nucleic Acid Modification and Labelling
Therminator DNA Polymerase is a 9°N™ DNA Polymerase variant with an enhanced ability to incorporate modified substrates such as dideoxynucleotides, ribonucleotides and acyclonucleotides.