TaqI-v2, New England Biolabs
Supplier: New England Biolabs (NEB)
R0149L R0149M R0149S R0149T
CA101228-810EA 438 CAD
CA101228-810 CA101228-812 CA101228-814 CA101228-816
TaqI-v2, New England Biolabs
Nucleic Acid Reagents Restriction Enzymes
An E.coli strain that carries a TaqαI overproducing plasmid (F. Barany using an NEB clone). TaqαI has a two amino acid replacement at its amino terminus. This allows for a higher level of expression without interfering with its catalytic properites.