T7 RNA Polymerase, New England Biolabs
Supplier: New England Biolabs (NEB)
M0251L M0251S
CA101228-278EA 441 CAD
CA101228-278 CA101228-280
T7 RNA Polymerase, New England Biolabs
Nucleic Acid Reagents RNA Synthesis and Transcription
Bacteriophage T7 RNA Polymerase is a DNA-dependent RNA polymerase that is highly specific for the T7 phage promoters
  • Fast, streamlined workflow and novel master mixes
  • High yield preparation of high quality libraries
  • Functionally validated by preparation of a library
The 99 KD enzyme catalyzes in vitro RNA synthesis from a cloned DNA sequence under the T7 promoters. RNA produced using the T7 RNA Polymerase is suitable for many applications in research and biotechnology.
Certifications: In addition to the extensive QCs on individual kit components, all NEBNext kits for Illumina are functionally validated by preparation of a library, followed by Illumina sequencing.