T7 Endonuclease I, New England Biolabs
Supplier: New England Biolabs (NEB)
M0302L M0302S
CA101228-380EA 467 CAD
CA101228-380 CA101228-382
T7 Endonuclease I, New England Biolabs
Enzymes
T7 Endonuclease I recognizes and cleaves non-perfectly matched DNA, cruciform DNA structures, Holliday structures or junctions, heteroduplex DNA and more slowly, nicked double-stranded DNA
The cleavage site is at the first, second or third phosphodiester bond that is 5´ to the mismatch. The protein is the product of T7 gene 3.
Delivery information: Supplied with 10X Reaction Buffer, NEBuffer 2