T7 DNA Polymerase (Unmodified), New England Biolabs
Supplier: New England Biolabs (NEB)
M0274S M0274L M0274S
200064-432EA 129.56 CAD
200064-432 CA200064-430 CA200064-432
T7 DNA Polymerase (Unmodified), New England Biolabs
Nucleic Acid Reagents Nucleic Acid Modification and Labelling
T7 DNA Polymerase catalyzes the replication of T7 phage DNA during infection
The protein dimer has two catalytic activities: DNA polymerase activity and strong 3´→ 5´ exonuclease. The high fidelity and rapid extension rate of the enzyme make it particularly useful in copying long stretches of DNA template.