T4 RNA Ligase 1 (ssRNA Ligase), New England Biolabs
Supplier: New England Biolabs (NEB)
M0204L M0204S
CA101228-188EA 456 CAD
CA101228-188 CA101228-190
T4 RNA Ligase 1 (ssRNA Ligase), New England Biolabs
Nucleic Acid Reagents Cloning Kits and Ligation
T4 RNA Ligase 1 catalyzes the ligation of a 5' phosphoryl-terminated nucleic acid donor to a 3' hydroxyl-terminated nucleic acid acceptor through the formation of a 3'→5' phosphodiester bond, with hydrolysis of ATP to AMP and PPi
Substrates include single-stranded RNA and DNA as well as dinucleoside pyrophosphates.