T4 Polynucleotide Kinase, New England Biolabs
Supplier: New England Biolabs (NEB)
M0201L M0201S
CA101228-172EA 359 CAD
CA101228-172 CA101228-174
T4 Polynucleotide Kinase, New England Biolabs
Enzymes
T4 Polynucleotide Kinase catalyzes the transfer and exchange of Pi from the γ position of ATP to the 5´ hydroxyl terminus of polynucleotides (double- and single-stranded DNA and RNA), as well as nucleoside 3´ monophosphates
The T4 Polynucleotide Kinase also catalyzes the removal of 3´ phosphoryl groups from 3´ phosphoryl polynucleotides, deoxy­nucleoside 3´ monophosphates and deoxynucleoside 3´ diphosphates.