T4 Polynucleotide Kinase, New England Biolabs
Supplier: New England Biolabs (NEB)
M0236L
M0236S
CA101417-956EA
721
CAD
CA101417-956
CA101417-958
T4 Polynucleotide Kinase, New England Biolabs
Enzymes
T4 Polynucleotide Kinase catalyzes the transfer and exchange of Pi from the γ position of ATP to the 5´ -hydroxyl terminus of double- and single-stranded DNA and RNA, as well as nucleoside 3´-monophosphates
This modified version exhibits full kinase activity with no 3´ phosphatase activity.