T4 Phage Beta-Glucosyltransferase, New England Biolabs
Supplier: New England Biolabs (NEB)
M0357L M0357S
CA101640-964EA 622 CAD
CA101640-964 CA101640-966
T4 Phage Beta-Glucosyltransferase, New England Biolabs
Enzymes
T4 Phage β-glucosyltransferase specifically transfers the glucose moiety of uridine diphosphoglucose (UDP-Glc) to the 5-hydroxymethylcytosine (5-hmC) residues in double-stranded DNA, making beta-glucosyl-5-hydroxymethylcytosine.